siRNA Details
| Gene Symbol: Rergl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191979.1 | NP_001178908.1 | 4 | 345 | ||
| XM_008763418.1 | XP_008761640.1 | 3 | 341 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rergl| Gene Symbol: Rergl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191979.1 | NP_001178908.1 | 4 | 345 | ||
| XM_008763418.1 | XP_008761640.1 | 3 | 341 | |||