siRNA Details
| Gene Symbol: RGD1561551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_575715.5 | XP_575715.1 | 5 | 1783 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1561551| Gene Symbol: RGD1561551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_575715.5 | XP_575715.1 | 5 | 1783 | ||