siRNA Details
| Gene Symbol: Lrmp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763454.1 | XP_008761676.1 | 16, 17 | 2343 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrmp| Gene Symbol: Lrmp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763454.1 | XP_008761676.1 | 16, 17 | 2343 | ||