siRNA Details
| Gene Symbol: Lmntd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191980.1 | NP_001178909.1 | 2 | 259 | ||
| XM_008763419.1 | XP_008761641.1 | 3 | 1578 | |||
| XM_008763420.1 | XP_008761642.1 | 2 | 143 | |||
| XM_008763421.1 | XP_008761643.1 | 1 | 187 | |||
| Gene Symbol: LOC100912509 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237675.2 | XP_006237737.1 | 1 | 281 | ||

Lmntd1
Genomic Map