siRNA Details
| Gene Symbol: Rep15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109256.1 | NP_001102726.1 | 1 | 620 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rep15| Gene Symbol: Rep15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109256.1 | NP_001102726.1 | 1 | 620 | ||