siRNA Details
| Gene Symbol: RGD1561849 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109260.1 | NP_001102730.1 | 6 | 931 | ||
| GenBank mRNA | FQ211566.1 | - | 6 | 845 | ||
| FQ212089.1 | - | 6 | 855 | |||
| FQ212229.1 | - | 6 | 855 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1561849| Gene Symbol: RGD1561849 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109260.1 | NP_001102730.1 | 6 | 931 | ||
| GenBank mRNA | FQ211566.1 | - | 6 | 845 | ||
| FQ212089.1 | - | 6 | 855 | |||
| FQ212229.1 | - | 6 | 855 | |||