siRNA Details
Gene Symbol: RGD1561849 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109260.1 | NP_001102730.1 | 6 | 958 | ||
GenBank mRNA | FQ211566.1 | - | 6 | 872 | ||
FQ212089.1 | - | 6 | 882 | |||
FQ212229.1 | - | 6 | 882 |
