siRNA Details
| Gene Symbol: Fam110b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024341.1 | NP_001019512.1 | 1 | 1048 | ||
| XM_006237839.2 | XP_006237901.1 | 3 | 2341 | |||
| XM_006237840.2 | XP_006237902.1 | 4 | 1569 | |||
| XM_006237841.2 | XP_006237903.1 | 3 | 1482 | |||
| XM_006237843.2 | XP_006237905.1 | 2 | 6672 | |||
| XM_008763551.1 | XP_008761773.1 | 2 | 7446 | |||
| XM_008763552.1 | XP_008761774.1 | 3 | 5201 | |||
| XM_008763553.1 | XP_008761775.1 | 4 | 4900 | |||
| XM_008763554.1 | XP_008761776.1 | 4 | 5150 | |||
| XM_008763555.1 | XP_008761777.1 | 2 | 2254 | |||
| GenBank mRNA | BC091289.1 | - | 1 | 1048 | 7306431 | |

Fam110b
Genomic Map