siRNA Details
| Gene Symbol: Triqk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171802.1 | NP_001165273.1 | 4 | 388 | ||
| XM_006237906.2 | XP_006237968.1 | 4 | 399 | |||
| GenBank mRNA | BC089993.1 | - | 4 | 358 | 7319652 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Triqk| Gene Symbol: Triqk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171802.1 | NP_001165273.1 | 4 | 388 | ||
| XM_006237906.2 | XP_006237968.1 | 4 | 399 | |||
| GenBank mRNA | BC089993.1 | - | 4 | 358 | 7319652 | |