siRNA Details
| Gene Symbol: Dnai1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024342.1 | NP_001019513.1 | 8, 9 | 804 | ||
| GenBank mRNA | BC083662.1 | - | 8, 9 | 804 | 7135392 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnai1| Gene Symbol: Dnai1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024342.1 | NP_001019513.1 | 8, 9 | 804 | ||
| GenBank mRNA | BC083662.1 | - | 8, 9 | 804 | 7135392 | |