siRNA Details
Gene Symbol: RGD1559864 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238121.2 | XP_006238183.1 | 3 | 1207 | ||
XM_006238122.2 | XP_006238184.1 | 4 | 986 | |||
XM_006238123.2 | XP_006238185.1 | 3 | 853 | |||
XM_008763652.1 | XP_008761874.1 | 2 | 862 | |||
XM_008763653.1 | XP_008761875.1 | 3 | 751 | |||
GenBank mRNA | FQ211775.1 | - | 2 | 641 |
