siRNA Details
Gene Symbol: RGD1559864 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238121.2 | XP_006238183.1 | 4, 5 | 1291 | ||
XM_006238122.2 | XP_006238184.1 | 5, 6 | 1070 | |||
XM_006238123.2 | XP_006238185.1 | 4, 5 | 937 | |||
XM_008763652.1 | XP_008761874.1 | 3, 4 | 946 | |||
XM_008763653.1 | XP_008761875.1 | 4, 5 | 835 | |||
GenBank mRNA | FQ211775.1 | - | 3, 4 | 725 |
