siRNA Details
| Gene Symbol: RGD1559864 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238121.2 | XP_006238183.1 | 7, 8 | 1755 | ||
| XM_006238122.2 | XP_006238184.1 | 8, 9 | 1534 | |||
| XM_006238123.2 | XP_006238185.1 | 7, 8 | 1401 | |||
| XM_008763652.1 | XP_008761874.1 | 6, 7 | 1410 | |||
| XM_008763653.1 | XP_008761875.1 | 7, 8 | 1299 | |||
| GenBank mRNA | FQ211775.1 | - | 6, 7 | 1189 | ||

RGD1559864
Genomic Map