siRNA Details
Gene Symbol: Sit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024344.1 | NP_001019515.1 | 1 | 130 | ||
GenBank mRNA | BC088199.1 | - | 1 | 130 | 7377221 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024344.1 | NP_001019515.1 | 1 | 130 | ||
GenBank mRNA | BC088199.1 | - | 1 | 130 | 7377221 |