siRNA Details
Gene Symbol: Epb41l4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127565.1 | NP_001121037.1 | 12 | 1243 | ||
GenBank mRNA | BC166749.1 | - | 12 | 1243 | 5599589 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Epb41l4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127565.1 | NP_001121037.1 | 12 | 1243 | ||
GenBank mRNA | BC166749.1 | - | 12 | 1243 | 5599589 |