siRNA Details
Gene Symbol: Rnf183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238243.2 | XP_006238305.1 | 6 | 1473 | ||
XM_008763806.1 | XP_008762028.1 | 5 | 1588 | |||
XM_008763807.1 | XP_008762029.1 | 3 | 766 | |||
XM_008763808.1 | XP_008762030.1 | 5 | 1251 | |||
XM_008763809.1 | XP_008762031.1 | 4 | 1078 | |||
XM_008763810.1 | XP_008762032.1 | 2 | 578 |
