siRNA Details
| Gene Symbol: LOC500475 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024346.1 | NP_001019517.1 | 1 | 416 | ||
| XM_008763797.1 | XP_008762019.1 | 1 | 629 | |||
| GenBank mRNA | BC079053.1 | - | 1 | 416 | 7115405 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC500475| Gene Symbol: LOC500475 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024346.1 | NP_001019517.1 | 1 | 416 | ||
| XM_008763797.1 | XP_008762019.1 | 1 | 629 | |||
| GenBank mRNA | BC079053.1 | - | 1 | 416 | 7115405 | |