siRNA Details
| Gene Symbol: Eqtn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039345.1 | NP_001034434.1 | 4 | 426 | ||
| XM_008763895.1 | XP_008762117.1 | 4 | 613 | |||
| XM_008763896.1 | XP_008762118.1 | 4 | 613 | |||
| GenBank mRNA | DQ336136.1 | - | 4 | 426 | ||

Eqtn
Genomic Map