siRNA Details
| Gene Symbol: RGD1560146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134625.1 | NP_001128097.1 | 7, 8 | 1112 | ||
| XM_006238420.2 | XP_006238482.1 | 8, 9 | 1190 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1560146| Gene Symbol: RGD1560146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134625.1 | NP_001128097.1 | 7, 8 | 1112 | ||
| XM_006238420.2 | XP_006238482.1 | 8, 9 | 1190 | |||