siRNA Details
| Gene Symbol: Lurap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109263.1 | NP_001102733.1 | 2 | 692 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lurap1| Gene Symbol: Lurap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109263.1 | NP_001102733.1 | 2 | 692 | ||