siRNA Details
| Gene Symbol: RGD1563015 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134627.2 | NP_001128099.2 | 6 | 624 | ||
| XM_008764055.1 | XP_008762277.1 | 6 | 793 | |||
| XM_008764056.1 | XP_008762278.1 | 7 | 1387 | |||
| XM_008764057.1 | XP_008762279.1 | 5 | 721 | |||

RGD1563015
Genomic Map