siRNA Details
| Gene Symbol: S100pbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134628.1 | NP_001128100.1 | 5 | 1167 | ||
| XM_006238956.2 | XP_006239018.1 | 5 | 1134 | |||
| XM_006238957.2 | XP_006239019.1 | 5 | 1438 | |||
| XM_006238958.2 | XP_006239020.1 | 5 | 1150 | |||
| XM_006238959.2 | XP_006239021.1 | 4 | 1020 | |||
| XM_006238960.1 | XP_006239022.1 | 4 | 1079 | |||
| XM_006238961.2 | XP_006239023.1 | 6 | 1008 | |||
| XM_006238962.2 | XP_006239024.1 | 5 | 1133 | |||
| XM_006238963.2 | XP_006239025.1 | 6 | 904 | |||
| XM_008764162.1 | XP_008762384.1 | 5 | 1134 | |||
| XM_008764163.1 | XP_008762385.1 | 5 | 1133 | |||
| XM_008764164.1 | XP_008762386.1 | 5 | 1133 | |||
| Gene Symbol: LOC498829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY325193.1 | - | 3 | 146 | ||

S100pbp
Genomic Map