siRNA Details
| Gene Symbol: RGD1561149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134629.1 | NP_001128101.1 | 6 | 3128 | ||
| XM_006238964.2 | XP_006239026.1 | 6 | 7502 | |||
| XM_006238965.2 | XP_006239027.1 | 6 | 7502 | |||
| XM_006238966.2 | XP_006239028.1 | 6 | 4961 | |||

RGD1561149
Genomic Map