siRNA Details
| Gene Symbol: Zbtb8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109265.1 | NP_001102735.1 | 2 | 1041 | ||
| XM_006238967.1 | XP_006239029.1 | 3 | 1103 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb8b| Gene Symbol: Zbtb8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109265.1 | NP_001102735.1 | 2 | 1041 | ||
| XM_006238967.1 | XP_006239029.1 | 3 | 1103 | |||