siRNA Details
| Gene Symbol: Zcchc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109267.1 | NP_001102737.1 | 4, 5 | 353 | ||
| XM_006238968.2 | XP_006239030.1 | 4, 5 | 373 | |||
| XM_006238969.2 | XP_006239031.1 | 3, 4 | 309 | |||
| XM_006238970.2 | XP_006239032.1 | 2, 3 | 188 | |||

Zcchc17
Genomic Map