siRNA Details
| Gene Symbol: Szrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114599.2 | NP_001108071.1 | 4 | 461 | ||
| XM_006239264.2 | XP_006239326.1 | 4 | 489 | |||
| GenBank mRNA | BC083784.1 | - | 5 | 543 | ||
| BP502965.1 | - | 4 | 448 | |||
| FQ213371.1 | - | 5 | 562 | |||

Szrd1
Genomic Map