siRNA Details
| Gene Symbol: Fbxo44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191576.1 | NP_001178505.1 | 2, 3 | 567 | ||
| XM_006239392.1 | XP_006239454.1 | 2, 3 | 589 | |||
| XM_006239393.1 | XP_006239455.1 | 2, 3 | 589 | |||
| XM_006239395.2 | XP_006239457.1 | 2, 3 | 370 | |||
| XM_006239396.2 | XP_006239458.2 | 2, 3 | 324 | |||
| XM_006239398.2 | XP_006239460.2 | 2, 3 | 324 | |||
| XM_006239399.2 | XP_006239461.1 | 2, 3 | 589 | |||
| GenBank mRNA | FQ213337.1 | - | 2, 3 | 296 | ||

Fbxo44
Genomic Map