siRNA Details
| Gene Symbol: Fbxo44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191576.1 | NP_001178505.1 | 4 | 745 | ||
| XM_006239392.1 | XP_006239454.1 | 4 | 767 | |||
| XM_006239395.2 | XP_006239457.1 | 4 | 548 | |||
| XM_006239396.2 | XP_006239458.2 | 4 | 502 | |||
| XM_006239399.2 | XP_006239461.1 | 4 | 738 | |||
| GenBank mRNA | FQ213337.1 | - | 4 | 445 | ||

Fbxo44
Genomic Map