siRNA Details
| Gene Symbol: LOC500594 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047958.1 | NP_001041423.1 | 4, 5 | 653 | ||
| GenBank mRNA | AY539913.1 | - | 4, 5 | 653 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC500594| Gene Symbol: LOC500594 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047958.1 | NP_001041423.1 | 4, 5 | 653 | ||
| GenBank mRNA | AY539913.1 | - | 4, 5 | 653 | ||