siRNA Details
| Gene Symbol: RGD1562146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AW916959.1 | - | 1 | 192 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1562146| Gene Symbol: RGD1562146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AW916959.1 | - | 1 | 192 | ||