siRNA Details
| Gene Symbol: Socs5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109274.1 | NP_001102744.1 | 2 | 1888 | ||
| XM_006239718.2 | XP_006239780.1 | 2 | 1591 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Socs5| Gene Symbol: Socs5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109274.1 | NP_001102744.1 | 2 | 1888 | ||
| XM_006239718.2 | XP_006239780.1 | 2 | 1591 | |||