siRNA Details
| Gene Symbol: RGD1563701 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750129.3 | XP_003750177.1 | 2 | 698 | ||
| XM_006239866.1 | XP_006239928.1 | 2 | 664 | |||
| XM_006239868.2 | XP_006239930.1 | 2 | 770 | |||
| XM_008764556.1 | XP_008762778.1 | 2 | 612 | |||

RGD1563701
Genomic Map