siRNA Details
| Gene Symbol: Fscb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271107.1 | NP_001258036.1 | 1 | 856 | ||
| GenBank mRNA | BC098054.1 | - | 1 | 248 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fscb| Gene Symbol: Fscb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271107.1 | NP_001258036.1 | 1 | 856 | ||
| GenBank mRNA | BC098054.1 | - | 1 | 248 | ||