siRNA Details
| Gene Symbol: Dact1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729635.3 | XP_002729681.1 | 4 | 925 | ||
| XM_576044.5 | XP_576044.1 | 4 | 1036 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dact1| Gene Symbol: Dact1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729635.3 | XP_002729681.1 | 4 | 925 | ||
| XM_576044.5 | XP_576044.1 | 4 | 1036 | |||