siRNA Details
| Gene Symbol: Ccdc175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164400.1 | NP_001157872.1 | 13 | 1576 | ||
| XM_008764718.1 | XP_008762940.1 | 13 | 1619 | |||
| GenBank mRNA | BC087160.1 | - | 13 | 1576 | 7134746 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc175| Gene Symbol: Ccdc175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164400.1 | NP_001157872.1 | 13 | 1576 | ||
| XM_008764718.1 | XP_008762940.1 | 13 | 1619 | |||
| GenBank mRNA | BC087160.1 | - | 13 | 1576 | 7134746 | |