siRNA Details
| Gene Symbol: Adam4l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109277.1 | NP_001102747.1 | 1 | 904 | ||
| GenBank mRNA | BC079009.1 | - | 1 | 882 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adam4l1| Gene Symbol: Adam4l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109277.1 | NP_001102747.1 | 1 | 904 | ||
| GenBank mRNA | BC079009.1 | - | 1 | 882 | ||