siRNA Details
| Gene Symbol: Ccdc176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240348.2 | XP_006240410.1 | 8 | 1180 | ||
| XM_008764829.1 | XP_008763051.1 | 8 | 1180 | |||
| XM_008764830.1 | XP_008763052.1 | 8 | 1053 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc176| Gene Symbol: Ccdc176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240348.2 | XP_006240410.1 | 8 | 1180 | ||
| XM_008764829.1 | XP_008763051.1 | 8 | 1180 | |||
| XM_008764830.1 | XP_008763052.1 | 8 | 1053 | |||