siRNA Details
| Gene Symbol: LOC500702 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047960.1 | NP_001041425.1 | 1, 2 | 111 | ||
| GenBank mRNA | AY383706.1 | - | 1, 2 | 111 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC500702| Gene Symbol: LOC500702 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047960.1 | NP_001041425.1 | 1, 2 | 111 | ||
| GenBank mRNA | AY383706.1 | - | 1, 2 | 111 | ||