siRNA Details
| Gene Symbol: Efcab11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024350.1 | NP_001019521.1 | 2 | 195 | ||
| XM_006240446.2 | XP_006240508.2 | 2 | 297 | |||
| GenBank mRNA | BC079408.1 | - | 2 | 195 | 7113005 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Efcab11| Gene Symbol: Efcab11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024350.1 | NP_001019521.1 | 2 | 195 | ||
| XM_006240446.2 | XP_006240508.2 | 2 | 297 | |||
| GenBank mRNA | BC079408.1 | - | 2 | 195 | 7113005 | |