siRNA Details
| Gene Symbol: Zfp839 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240613.2 | XP_006240675.1 | 2 | 1781 | ||
| XM_006240614.2 | XP_006240676.1 | 2 | 1781 | |||
| XM_006240615.2 | XP_006240677.1 | 2 | 1276 | |||
| XM_006240616.2 | XP_006240678.2 | 2 | 1985 | |||
| XM_008764973.1 | XP_008763195.1 | 2 | 1986 | |||

Zfp839
Genomic Map