siRNA Details
| Gene Symbol: RGD1560608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109280.1 | NP_001102750.1 | 2, 3 | 169 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1560608| Gene Symbol: RGD1560608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109280.1 | NP_001102750.1 | 2, 3 | 169 | ||