siRNA Details
| Gene Symbol: Cep170b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729727.4 | XP_002729773.2 | 12 | 3300 | ||
| XM_008764990.1 | XP_008763212.1 | 12 | 4113 | |||
| XM_008764991.1 | XP_008763213.1 | 13 | 3952 | |||
| XM_576105.5 | XP_576105.4 | 12 | 3300 | |||

Cep170b
Genomic Map