siRNA Details
| Gene Symbol: Cdca4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037214.1 | NP_001032291.1 | 2 | 550 | ||
| XM_008764947.1 | XP_008763169.1 | 3 | 673 | |||
| XM_008764948.1 | XP_008763170.1 | 2 | 414 | |||
| GenBank mRNA | BC107668.1 | - | 2 | 550 | 7937865 | |

Cdca4
Genomic Map