siRNA Details
| Gene Symbol: Apof | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024351.1 | NP_001019522.1 | 2 | 914 | ||
| GenBank mRNA | AY325229.1 | - | 663 | |||
| BC088170.1 | - | 2 | 914 | 7322324 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Apof| Gene Symbol: Apof | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024351.1 | NP_001019522.1 | 2 | 914 | ||
| GenBank mRNA | AY325229.1 | - | 663 | |||
| BC088170.1 | - | 2 | 914 | 7322324 | ||