siRNA Details
Gene Symbol: Btbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241037.2 | XP_006241099.2 | 6 | 1005 | ||
XM_008765152.1 | XP_008763374.1 | 6 | 1023 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Btbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241037.2 | XP_006241099.2 | 6 | 1005 | ||
XM_008765152.1 | XP_008763374.1 | 6 | 1023 |