siRNA Details
| Gene Symbol: Misp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109284.1 | NP_001102754.1 | 2 | 1239 | ||
| XM_008765176.1 | XP_008763398.1 | 3 | 3007 | |||
| XM_008765177.1 | XP_008763399.1 | 2 | 2836 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Misp| Gene Symbol: Misp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109284.1 | NP_001102754.1 | 2 | 1239 | ||
| XM_008765176.1 | XP_008763398.1 | 3 | 3007 | |||
| XM_008765177.1 | XP_008763399.1 | 2 | 2836 | |||