siRNA Details
Gene Symbol: Slc5a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191987.1 | NP_001178916.1 | 12 | 1460 | ||
XM_006241232.2 | XP_006241294.1 | 12 | 3118 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc5a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191987.1 | NP_001178916.1 | 12 | 1460 | ||
XM_006241232.2 | XP_006241294.1 | 12 | 3118 |