siRNA Details
| Gene Symbol: Ccdc38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241271.2 | XP_006241333.1 | 15, 16 | 1592 | ||
| XM_008765310.1 | XP_008763532.1 | 14, 15 | 1322 | |||
| XM_008765311.1 | XP_008763533.1 | 15, 16 | 1604 | |||

Ccdc38
Genomic Map