siRNA Details
| Gene Symbol: Galnt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025053.2 | NP_001020224.2 | 1 | 988 | ||
| XM_006241297.2 | XP_006241359.1 | 1 | 987 | |||
| GenBank mRNA | AB040675.1 | - | 1 | 742 | ||
| BC105818.1 | - | 1 | 988 | 7368805 | ||

Galnt4
Genomic Map