siRNA Details
| Gene Symbol: Bbs10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109286.1 | NP_001102756.1 | 2 | 2067 | ||
| XM_006241323.2 | XP_006241385.1 | 2 | 2033 | |||
| GenBank mRNA | BC160915.1 | - | 2 | 2059 | 7457522 | |
| FQ211675.1 | - | 2 | 2049 | |||

Bbs10
Genomic Map